Product Details

SNP ID
rs142193912
Assay Type
Functionally tested
NCBI dbSNP Submissions
16
Location
Chr.1:162855082 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCACATGCCATTCCACTATCACTAG[C/T]TGGGCTGGAACTAGGACCTTGTTCT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCDC190 PubMed Links

Gene Details

Gene
CCDC190
Gene Name
coiled-coil domain containing 190
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178550.4 3385 Missense Mutation ACT,GCT T198A NP_848645.3
XM_005245125.3 3385 Missense Mutation ACT,GCT T209A XP_005245182.1
XM_006711289.3 3385 Missense Mutation ACT,GCT T197A XP_006711352.1
XM_011509477.2 3385 Missense Mutation ACT,GCT T210A XP_011507779.1

View Full Product Details