Product Details

SNP ID
rs142228396
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:145936288 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATAAAAAAAGGCTGTGGAAAGCCT[G/A]GGGAGTTATGTGGGTGGATGCCTAG
Phenotype
MIM: 605313
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
LIX1L PubMed Links

Gene Details

Gene
LIX1L
Gene Name
limb and CNS expressed 1 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153713.2 1128 UTR 3 NP_714924.1
XM_017000316.1 1128 UTR 3 XP_016855805.1
Gene
RBM8A
Gene Name
RNA binding motif protein 8A
There are no transcripts associated with this gene.

View Full Product Details