Product Details

SNP ID
rs142344117
Assay Type
Functionally tested
NCBI dbSNP Submissions
9
Location
Chr.1:109066211 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTATAAGGATTCTGGTCATCCCCA[A/T]AGCCATACATCATACATCGCACTGT
Phenotype
MIM: 600774
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
TAF13 PubMed Links

Gene Details

Gene
TAF13
Gene Name
TATA-box binding protein associated factor 13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005645.3 183 Missense Mutation TAT,TTT Y43F NP_005636.1

View Full Product Details