Product Details

SNP ID
rs142553340
Assay Type
Functionally tested
NCBI dbSNP Submissions
6
Location
Chr.1:203307348 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTTGTACGAGGAGGCCCCACTGGC[C/T]GCCTCCTGTGGGCTCCTCACCTGCA
Phenotype
MIM: 601597
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BTG2 PubMed Links

Gene Details

Gene
BTG2
Gene Name
BTG anti-proliferation factor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006763.2 458 Silent Mutation GCC,GCT A129A NP_006754.1
Gene
LINC01136
Gene Name
long intergenic non-protein coding RNA 1136
There are no transcripts associated with this gene.

View Full Product Details