Product Details

SNP ID
rs142584447
Assay Type
Functionally tested
NCBI dbSNP Submissions
10
Location
Chr.1:1203888 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCCGCCCCTTCTCCTCTGCCGATC[A/G]CTCGCCCCGCTCTTCCTCGGGGAAC
Phenotype
MIM: 603905 MIM: 600315
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TNFRSF18 PubMed Links

Gene Details

Gene
TNFRSF18
Gene Name
TNF receptor superfamily member 18
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004195.2 799 Nonsense Mutation CGA,TGA R228* NP_004186.1
NM_148901.1 799 Silent Mutation AGC,AGT S157S NP_683699.1
NM_148902.1 799 Nonsense Mutation CGA,TGA R221* NP_683700.1
XM_017002722.1 799 Silent Mutation AGC,AGT S249S XP_016858211.1
Gene
TNFRSF4
Gene Name
TNF receptor superfamily member 4
There are no transcripts associated with this gene.

Gene
TTLL10
Gene Name
tubulin tyrosine ligase like 10
There are no transcripts associated with this gene.

View Full Product Details