Product Details

SNP ID
rs142622304
Assay Type
Functionally tested
NCBI dbSNP Submissions
8
Location
Chr.1:230322646 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCATGCAAAGAGCCGTGACTGAGG[C/G]CGGGGTCAGCGTCGGGGGTGGAGAG
Phenotype
MIM: 616791
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
PGBD5 PubMed Links

Gene Details

Gene
PGBD5
Gene Name
piggyBac transposable element derived 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001258311.1 3423 UTR 3 NP_001245240.1
XM_017002330.1 3423 UTR 3 XP_016857819.1

View Full Product Details