Product Details
- SNP ID
-
rs142661836
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
13
- Location
-
Chr.1:1180196 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCCCCTGGGCTCCTGAACAGCCACC[A/G]GCCTGCAGACTCGGATGACACTAAC
- Phenotype
-
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
TTLL10
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs111751804] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- TTLL10
- Gene Name
- tubulin tyrosine ligase like 10
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001130045.1 |
672 |
Missense Mutation |
CAG,CGG |
Q121R |
NP_001123517.1 |
NM_153254.2 |
672 |
Missense Mutation |
CAG,CGG |
Q48R |
NP_694986.2 |
XM_005244738.1 |
672 |
Missense Mutation |
CAG,CGG |
Q48R |
XP_005244795.1 |
XM_011541177.2 |
672 |
Missense Mutation |
CAG,CGG |
Q121R |
XP_011539479.1 |
XM_017000906.1 |
672 |
Missense Mutation |
CAG,CGG |
Q94R |
XP_016856395.1 |
XM_017000907.1 |
672 |
Missense Mutation |
CAG,CGG |
Q121R |
XP_016856396.1 |
XM_017000908.1 |
672 |
Missense Mutation |
CAG,CGG |
Q121R |
XP_016856397.1 |
XM_017000909.1 |
672 |
Missense Mutation |
CAG,CGG |
Q121R |
XP_016856398.1 |
XM_017000910.1 |
672 |
Missense Mutation |
CAG,CGG |
Q121R |
XP_016856399.1 |
XM_017000911.1 |
672 |
Missense Mutation |
CAG,CGG |
Q121R |
XP_016856400.1 |
XM_017000912.1 |
672 |
Missense Mutation |
CAG,CGG |
Q121R |
XP_016856401.1 |
- Gene
- TTLL10-AS1
- Gene Name
- TTLL10 antisense RNA 1
There are no transcripts associated with this gene.
View Full Product Details