Product Details
- SNP ID
-
rs142722847
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
10
- Location
-
Chr.1:155197469 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AATCTGAGCAGCTGGGGACCTTGAG[A/C]TGCAGCCCGGGCTGGGCAACCGCCC
- Phenotype
-
MIM: 158340
MIM: 188062
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
MIR92B
PubMed Links
Gene Details
- Gene
- MIR92B
- Gene Name
- microRNA 92b
There are no transcripts associated with this gene.
- Gene
- MUC1
- Gene Name
- mucin 1, cell surface associated
There are no transcripts associated with this gene.
- Gene
- THBS3
- Gene Name
- thrombospondin 3
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001252607.1 |
2549 |
Missense Mutation |
CAG,CAT |
Q822H |
NP_001239536.1 |
NM_001252608.1 |
2549 |
Missense Mutation |
CAG,CAT |
Q711H |
NP_001239537.1 |
NM_007112.4 |
2549 |
Missense Mutation |
CAG,CAT |
Q831H |
NP_009043.1 |
XM_006711498.3 |
2549 |
Missense Mutation |
CAG,CAT |
Q888H |
XP_006711561.1 |
XM_011509926.2 |
2549 |
Missense Mutation |
CAG,CAT |
Q891H |
XP_011508228.1 |
XM_011509927.2 |
2549 |
Missense Mutation |
CAG,CAT |
Q882H |
XP_011508229.1 |
XM_011509928.2 |
2549 |
Missense Mutation |
CAG,CAT |
Q860H |
XP_011508230.1 |
XM_011509929.1 |
2549 |
Missense Mutation |
CAG,CAT |
Q834H |
XP_011508231.1 |
XM_011509930.2 |
2549 |
Missense Mutation |
CAG,CAT |
Q891H |
XP_011508232.1 |
XM_011509931.1 |
2549 |
Missense Mutation |
CAG,CAT |
Q796H |
XP_011508233.1 |
XM_011509932.2 |
2549 |
Missense Mutation |
CAG,CAT |
Q796H |
XP_011508234.1 |
XM_011509933.1 |
2549 |
Missense Mutation |
CAG,CAT |
Q796H |
XP_011508235.1 |
XM_011509934.1 |
2549 |
Missense Mutation |
CAG,CAT |
Q655H |
XP_011508236.1 |
XM_011509935.1 |
2549 |
Missense Mutation |
CAG,CAT |
Q655H |
XP_011508237.1 |
XM_011509936.1 |
2549 |
Missense Mutation |
CAG,CAT |
Q579H |
XP_011508238.1 |
XM_017002206.1 |
2549 |
Missense Mutation |
CAG,CAT |
Q363H |
XP_016857695.1 |
View Full Product Details