Product Details

SNP ID
rs142744958
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:248349184 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACTACCCTGTGATCGTGAACTCTC[A/G]AATCTGCATCCAGATGACACTGGCC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR14C36 PubMed Links

Gene Details

Gene
OR14C36
Gene Name
olfactory receptor family 14 subfamily C member 36
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001918.1 410 Missense Mutation CAA,CGA Q137R NP_001001918.1

View Full Product Details