Product Details
- SNP ID
-
rs142759538
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
3
- Location
-
Chr.1:20744963 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTTGGCAGGCGTTTTGGCCTTAGGA[A/G]GTGCTTTCTTAGGCAAGGGCCTAGC
- Phenotype
-
MIM: 616072
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
HP1BP3
PubMed Links
Gene Details
- Gene
- HP1BP3
- Gene Name
- heterochromatin protein 1 binding protein 3
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_016287.3 |
2072 |
Missense Mutation |
CCT,CTT |
P499L |
NP_057371.2 |
XM_005245875.4 |
2072 |
Missense Mutation |
CCT,CTT |
P499L |
XP_005245932.1 |
XM_005245876.3 |
2072 |
Missense Mutation |
CCT,CTT |
P499L |
XP_005245933.1 |
XM_005245877.4 |
2072 |
Missense Mutation |
CCT,CTT |
P499L |
XP_005245934.1 |
XM_005245878.4 |
2072 |
Missense Mutation |
CCT,CTT |
P499L |
XP_005245935.1 |
XM_005245879.4 |
2072 |
Missense Mutation |
CCT,CTT |
P461L |
XP_005245936.1 |
XM_011541532.1 |
2072 |
Missense Mutation |
CCT,CTT |
P461L |
XP_011539834.1 |
XM_011541533.1 |
2072 |
Missense Mutation |
CCT,CTT |
P461L |
XP_011539835.1 |
XM_011541534.2 |
2072 |
Missense Mutation |
CCT,CTT |
P461L |
XP_011539836.1 |
XM_017001393.1 |
2072 |
Missense Mutation |
CCT,CTT |
P499L |
XP_016856882.1 |
XM_017001394.1 |
2072 |
Missense Mutation |
CCT,CTT |
P461L |
XP_016856883.1 |
View Full Product Details