Product Details

SNP ID
rs142849555
Assay Type
Functionally tested
NCBI dbSNP Submissions
18
Location
Chr.1:23750898 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGAAGAGCAATTCCCGAAAGCGCC[A/C]TCGGGATGCCCTGCAGAAGGAGGAG
Phenotype
MIM: 600786
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
TCEB3 PubMed Links

Gene Details

Gene
TCEB3
Gene Name
transcription elongation factor B subunit 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003198.2 431 Missense Mutation CAT,CCT H124P NP_003189.2
Gene
TCEB3-AS1
Gene Name
TCEB3 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details