Product Details

SNP ID
rs142850057
Assay Type
Functionally tested
NCBI dbSNP Submissions
13
Location
Chr.1:178548264 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTGAACTGGACTCCCCAGCCTCAT[A/G]TTCCTCTGCAGGATGTAAAAGGTAA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C1orf220 PubMed Links

Gene Details

Gene
C1orf220
Gene Name
chromosome 1 open reading frame 220
There are no transcripts associated with this gene.

View Full Product Details