Product Details
- SNP ID
-
rs142920998
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
6
- Location
-
Chr.1:25553955 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAGAACTGGACAGACACGCGGGAGA[A/C]GCTGCTGGAGGGGATGCTGTTCAGC
- Phenotype
-
MIM: 605747
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
LDLRAP1
PubMed Links
Gene Details
- Gene
- LDLRAP1
- Gene Name
- low density lipoprotein receptor adaptor protein 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_015627.2 |
1178 |
Missense Mutation |
AAG,ACG |
K41T |
NP_056442.2 |
XM_006710559.3 |
1178 |
Missense Mutation |
AAG,ACG |
K41T |
XP_006710622.1 |
XM_006710560.3 |
1178 |
Missense Mutation |
AAG,ACG |
K41T |
XP_006710623.1 |
XM_006710561.3 |
1178 |
Missense Mutation |
AAG,ACG |
K41T |
XP_006710624.1 |
XM_011541209.2 |
1178 |
Missense Mutation |
AAG,ACG |
K41T |
XP_011539511.1 |
XM_011541210.2 |
1178 |
Missense Mutation |
AAG,ACG |
K41T |
XP_011539512.1 |
XM_011541211.2 |
1178 |
Missense Mutation |
AAG,ACG |
K41T |
XP_011539513.1 |
XM_011541212.2 |
1178 |
Missense Mutation |
AAG,ACG |
K41T |
XP_011539514.1 |
XM_017000994.1 |
1178 |
Missense Mutation |
AAG,ACG |
K14T |
XP_016856483.1 |
XM_017000995.1 |
1178 |
Missense Mutation |
AAG,ACG |
K41T |
XP_016856484.1 |
View Full Product Details