Product Details

SNP ID
rs143023872
Assay Type
Functionally tested
NCBI dbSNP Submissions
19
Location
Chr.1:158466076 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTCATCTGCCCTTCCACTCCTCCA[A/G]CCAGCTCCATCACTTCTTCTGTGAC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR10K1 PubMed Links

Gene Details

Gene
OR10K1
Gene Name
olfactory receptor family 10 subfamily K member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004473.1 515 Missense Mutation AAC,AGC N172S NP_001004473.1

View Full Product Details