Product Details

SNP ID
rs143137206
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:159535204 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACGTCACTGAGAGAACAAGGTCTTT[C/T]TCTATTGAACTTTCTGACTTCGGCT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR10J5 PubMed Links

Gene Details

Gene
OR10J5
Gene Name
olfactory receptor family 10 subfamily J member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004469.1 804 Silent Mutation GAA,GAG E268E NP_001004469.1

View Full Product Details