Product Details

SNP ID
rs143195635
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:197088263 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAACTGACTAATACTTACAGAGGCT[C/G]TATTTGTTGTCTTCAGTAAAATAGC
Phenotype
MIM: 605481
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ASPM PubMed Links

Gene Details

Gene
ASPM
Gene Name
abnormal spindle microtubule assembly
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001206846.1 10411 Missense Mutation ACA,AGA T1800R NP_001193775.1
NM_018136.4 10411 Missense Mutation ACA,AGA T3385R NP_060606.3

View Full Product Details