Product Details

SNP ID
rs143197714
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:119915532 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTGTGACTGGGTGTTCGCTCAGCA[C/G]CATTTGAGGAAGCATAACTGTGCTG
Phenotype
MIM: 600275
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
NOTCH2 PubMed Links

Gene Details

Gene
NOTCH2
Gene Name
notch 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001200001.1 7487 Intron NP_001186930.1
NM_024408.3 7487 Missense Mutation GCT,GGT A2397G NP_077719.2
XM_005270901.3 7487 Missense Mutation GCT,GGT A2358G XP_005270958.1
XM_011541519.2 7487 Missense Mutation GCT,GGT A2393G XP_011539821.1
XM_011541520.2 7487 Missense Mutation GCT,GGT A2358G XP_011539822.1
XM_017001372.1 7487 Missense Mutation GCT,GGT A2381G XP_016856861.1
XM_017001373.1 7487 Missense Mutation GCT,GGT A2358G XP_016856862.1

View Full Product Details