Product Details

SNP ID
rs143236819
Assay Type
Functionally tested
NCBI dbSNP Submissions
10
Location
Chr.1:51116803 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAATCTGGCCAGTAGGTCTTTGTAT[A/G]TGATATTCTCTCTGTCTAGATATAC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C1orf185 PubMed Links

Gene Details

Gene
C1orf185
Gene Name
chromosome 1 open reading frame 185
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136508.1 Intron NP_001129980.1
XM_011541282.2 Intron XP_011539584.1

View Full Product Details