Product Details

SNP ID
rs143241495
Assay Type
Functionally tested
NCBI dbSNP Submissions
16
Location
Chr.1:159535583 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCAAAGGACCCACACACCAGCTGG[A/G]CACATAGTCCCTTGCTCATGATGAC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR10J5 PubMed Links

Gene Details

Gene
OR10J5
Gene Name
olfactory receptor family 10 subfamily J member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004469.1 425 Missense Mutation GCC,GTC A142V NP_001004469.1

View Full Product Details