Product Details
- SNP ID
-
rs143251974
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
14
- Location
-
Chr.1:158579013 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCTGAATATGATGGGGCTGAGGAAG[C/G]GGGTAATGACAGTATAAGGGACTGC
- Phenotype
-
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
OR10X1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs36102561] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- OR10X1
- Gene Name
- olfactory receptor family 10 subfamily X member 1 (gene/pseudogene)
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001004477.1 |
887 |
Missense Mutation |
CCC,CGC |
P296R |
NP_001004477.1 |
View Full Product Details