Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_152489.1 | 277 | Intron | NP_689702.1 | ||
XM_005270498.4 | 277 | UTR 5 | XP_005270555.1 | ||
XM_006710379.3 | 277 | Intron | XP_006710442.1 | ||
XM_011540764.2 | 277 | UTR 5 | XP_011539066.1 | ||
XM_017000379.1 | 277 | Intron | XP_016855868.1 | ||
XM_017000380.1 | 277 | Intron | XP_016855869.1 | ||
XM_017000381.1 | 277 | Intron | XP_016855870.1 | ||
XM_017000382.1 | 277 | Intron | XP_016855871.1 | ||
XM_017000383.1 | 277 | Intron | XP_016855872.1 | ||
XM_017000384.1 | 277 | Intron | XP_016855873.1 | ||
XM_017000385.1 | 277 | Intron | XP_016855874.1 |