Product Details

SNP ID
rs143391224
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:9553740 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTACTATCCTCAGGTTCATCTGGG[A/G]ACCATTAGTGGAGCTGGAATGGTGA
Phenotype
MIM: 610816
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SLC25A33 PubMed Links

Gene Details

Gene
SLC25A33
Gene Name
solute carrier family 25 member 33
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032315.2 181 Intron NP_115691.1
XM_005263503.3 181 Intron XP_005263560.1
XM_011542296.1 181 UTR 5 XP_011540598.1

View Full Product Details