Product Details
- SNP ID
-
rs143474721
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
16
- Location
-
Chr.1:153618972 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCCCCAATCAATCTCCAGTACTCAT[G/T]GAACTTGAGCTCCGAGTCCTGATTC
- Phenotype
-
MIM: 176940
MIM: 601989
MIM: 607986
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
S100A1
PubMed Links
Gene Details
- Gene
- S100A1
- Gene Name
- S100 calcium binding protein A1
There are no transcripts associated with this gene.
- Gene
- S100A13
- Gene Name
- S100 calcium binding protein A13
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001024210.1 |
548 |
Missense Mutation |
AAT,CAT |
N74H |
NP_001019381.1 |
NM_001024211.1 |
548 |
Missense Mutation |
AAT,CAT |
N74H |
NP_001019382.1 |
NM_001024212.1 |
548 |
Missense Mutation |
AAT,CAT |
N74H |
NP_001019383.1 |
NM_001024213.1 |
548 |
Missense Mutation |
AAT,CAT |
N74H |
NP_001019384.1 |
NM_005979.2 |
548 |
Missense Mutation |
AAT,CAT |
N74H |
NP_005970.1 |
XM_005245434.3 |
548 |
Missense Mutation |
AAT,CAT |
N74H |
XP_005245491.1 |
XM_011509862.2 |
548 |
Missense Mutation |
AAT,CAT |
N74H |
XP_011508164.1 |
XM_011509863.2 |
548 |
Missense Mutation |
AAT,CAT |
N74H |
XP_011508165.1 |
XM_011509864.1 |
548 |
Missense Mutation |
AAT,CAT |
N74H |
XP_011508166.1 |
XM_017002034.1 |
548 |
Missense Mutation |
AAT,CAT |
N74H |
XP_016857523.1 |
XM_017002035.1 |
548 |
Missense Mutation |
AAT,CAT |
N74H |
XP_016857524.1 |
XM_017002036.1 |
548 |
Missense Mutation |
AAT,CAT |
N74H |
XP_016857525.1 |
- Gene
- S100A14
- Gene Name
- S100 calcium binding protein A14
There are no transcripts associated with this gene.
- Gene
- S100A16
- Gene Name
- S100 calcium binding protein A16
There are no transcripts associated with this gene.
View Full Product Details