Product Details
- SNP ID
-
hCV166463826
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.1:146018454 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCTGAGAAGGCCATGGCCACATCCT[T/C]TGCTACCTTGATGGAGAAGGAGAGC
- Phenotype
-
MIM: 608374
- Polymorphism
- T/C, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
HFE2
PubMed Links
Gene Details
- Gene
- HFE2
- Gene Name
- hemochromatosis type 2 (juvenile)
There are no transcripts associated with this gene.
View Full Product Details