Product Details

SNP ID
rs143505871
Assay Type
Functionally tested
NCBI dbSNP Submissions
6
Location
Chr.1:1437140 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCGGCCAGCACAGCTCGGGTGAGG[C/T]TGCCCAGGATGCGGTGCGTGCTTCT
Phenotype
MIM: 611901
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC102724312 PubMed Links

Gene Details

Gene
LOC102724312
Gene Name
uncharacterized LOC102724312
There are no transcripts associated with this gene.

Gene
LOC107985729
Gene Name
uncharacterized LOC107985729
There are no transcripts associated with this gene.

Gene
TMEM88B
Gene Name
transmembrane protein 88B
There are no transcripts associated with this gene.

Gene
VWA1
Gene Name
von Willebrand factor A domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022834.4 513 Missense Mutation GCT,GTT A96V NP_073745.2
NM_199121.2 513 Intron NP_954572.2

View Full Product Details