Product Details

SNP ID
rs143565896
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:11273989 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTGCCGCTTGCCTCTACTACCTGT[C/T]CCCTCTGAAACTGGAGCACTTGGCT
Phenotype
MIM: 611632
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
UBIAD1 PubMed Links

Gene Details

Gene
UBIAD1
Gene Name
UbiA prenyltransferase domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013319.2 792 Missense Mutation TCC,TTC S153F NP_037451.1
XM_006710590.3 792 Missense Mutation TCC,TTC S153F XP_006710653.1
XM_011541304.2 792 Missense Mutation TCC,TTC S153F XP_011539606.1

View Full Product Details