Product Details
- SNP ID
-
rs143580706
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
4
- Location
-
Chr.1:27005978 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGGCAGCCAGGTGGGATAGGCGTGA[C/G]CCAGGAGAGGTTGCACGGGGGTCAC
- Phenotype
-
MIM: 616824
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
FAM46B
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs61737590] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- FAM46B
- Gene Name
- family with sequence similarity 46 member B
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_052943.3 |
1416 |
Missense Mutation |
GCT,GGT |
A415G |
NP_443175.2 |
- Gene
- TRNP1
- Gene Name
- TMF1-regulated nuclear protein 1
There are no transcripts associated with this gene.
View Full Product Details