Product Details
- SNP ID
-
rs143586869
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
12
- Location
-
Chr.1:165203199 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGAACACCATCAGCCACACAGAATT[A/T]CTTTCCAGATCTGAAAGACACTCTG
- Phenotype
-
MIM: 600298
- Polymorphism
- A/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
LMX1A
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs202181805] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- LMX1A
- Gene Name
- LIM homeobox transcription factor 1 alpha
View Full Product Details