Product Details

SNP ID
rs143664032
Assay Type
Functionally tested
NCBI dbSNP Submissions
7
Location
Chr.1:13475599 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATCTTCTCCCCCAACACACACCTC[A/G]ATCTGAGAGGCAGGTTATGCTCCAG
Phenotype
MIM: 615212
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LRRC38 PubMed Links

Gene Details

Gene
LRRC38
Gene Name
leucine rich repeat containing 38
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001010847.1 1286 UTR 3 NP_001010847.1

View Full Product Details