Product Details

SNP ID
rs143679565
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:15529116 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGAAAGGAAGAGAAATGGAAGTGA[C/G]AAAGTTGAGCATTTCCTGGCAGTTC
Phenotype
MIM: 602234
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CASP9 PubMed Links

Gene Details

Gene
CASP9
Gene Name
caspase 9
There are no transcripts associated with this gene.

Gene
DNAJC16
Gene Name
DnaJ heat shock protein family (Hsp40) member C16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001287811.1 175 Intron NP_001274740.1
NM_015291.3 175 Missense Mutation ACA,AGA T4R NP_056106.1

View Full Product Details