Product Details

SNP ID
rs143688605
Assay Type
Functionally tested
NCBI dbSNP Submissions
8
Location
Chr.1:200918463 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCTCTCATACTCACATCTAGCCAT[C/T]AACTATTCCTATTGGTTCTACCCTC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C1orf106 PubMed Links

Gene Details

Gene
C1orf106
Gene Name
chromosome 1 open reading frame 106
There are no transcripts associated with this gene.

Gene
MROH3P
Gene Name
maestro heat like repeat family member 3, pseudogene
There are no transcripts associated with this gene.

View Full Product Details