Product Details
- SNP ID
-
rs143708112
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
4
- Location
-
Chr.1:11668903 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GACTGGAAAATCTTCTACTTCCTAC[A/G]GAGCCTGCATAGGAACCTCCTGCGC
- Phenotype
-
MIM: 609111
MIM: 605647
MIM: 604094
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
FBXO44
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs7514198] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- FBXO44
- Gene Name
- F-box protein 44
There are no transcripts associated with this gene.
- Gene
- FBXO6
- Gene Name
- F-box protein 6
- Gene
- MAD2L2
- Gene Name
- MAD2 mitotic arrest deficient-like 2 (yeast)
There are no transcripts associated with this gene.
View Full Product Details