Product Details

SNP ID
rs143728385
Assay Type
Functionally tested
NCBI dbSNP Submissions
14
Location
Chr.1:248239595 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATACTTTCCTATTCCCATGTCCTT[C/T]GAGCCGTCATCCACATGGGCTCTGG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR2M4 PubMed Links

Gene Details

Gene
OR2M4
Gene Name
olfactory receptor family 2 subfamily M member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017504.1 667 Nonsense Mutation CGA,TGA R223* NP_059974.1

View Full Product Details