Product Details

SNP ID
rs143811617
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:156581708 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGAAGAGAAGGCACAGGGCAGGC[A/G]ACACGGCGACCAATGTTTCAATGTG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IQGAP3 PubMed Links

Gene Details

Gene
IQGAP3
Gene Name
IQ motif containing GTPase activating protein 3
There are no transcripts associated with this gene.

Gene
TTC24
Gene Name
tetratricopeptide repeat domain 24
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001105669.2 380 Missense Mutation CAA,CGA Q115R NP_001099139.2
XM_011509253.2 380 Intron XP_011507555.1
XM_011509254.2 380 Intron XP_011507556.1
XM_011509255.2 380 Intron XP_011507557.1
XM_017000497.1 380 Intron XP_016855986.1

View Full Product Details