Product Details
- SNP ID
-
rs143816164
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
9
- Location
-
Chr.1:223706628 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CACAACATGTCAGCTGCTCTGCTGG[A/G]CCTCTGTTTACTCATCCATTAAATG
- Phenotype
-
MIM: 114230
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CAPN2
PubMed Links
Gene Details
- Gene
- CAPN2
- Gene Name
- calpain 2
View Full Product Details