Product Details

SNP ID
rs143872295
Assay Type
Functionally tested
NCBI dbSNP Submissions
11
Location
Chr.1:94530477 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCTTCCTTTATGAAACATTCAGTG[A/G]GGAGTTCTCCTTCCAGCTCTGCCCC
Phenotype
MIM: 134390
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
F3 PubMed Links

Gene Details

Gene
F3
Gene Name
coagulation factor III, tissue factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001178096.1 1092 Silent Mutation CCC,CCT P237P NP_001171567.1
NM_001993.4 1092 Missense Mutation CCA,TCA P291S NP_001984.1

View Full Product Details