Product Details

SNP ID
rs143955130
Assay Type
Functionally tested
NCBI dbSNP Submissions
6
Location
Chr.1:84629002 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCCATTGATTTGCTTTGATTATTC[C/G]ACGATTTCAATCACTTTCCCAAAAC
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LINC01461 PubMed Links

Gene Details

Gene
LINC01461
Gene Name
long intergenic non-protein coding RNA 1461
There are no transcripts associated with this gene.

Gene
LINC01555
Gene Name
long intergenic non-protein coding RNA 1555
There are no transcripts associated with this gene.

View Full Product Details