Product Details

SNP ID
rs143991968
Assay Type
Functionally tested
NCBI dbSNP Submissions
13
Location
Chr.1:231370597 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGCTGGTTGTACTTCATGAGGGTT[A/G]CGACGGTCAGACCAGAAAAACAGCA
Phenotype
MIM: 606425
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EGLN1 PubMed Links

Gene Details

Gene
EGLN1
Gene Name
egl-9 family hypoxia inducible factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022051.2 1551 Silent Mutation CGC,CGT R371R NP_071334.1
XM_005273166.4 1551 Silent Mutation CGC,CGT R371R XP_005273223.1
XM_005273167.4 1551 Intron XP_005273224.1

View Full Product Details