Product Details

SNP ID
rs144041308
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:202335691 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGTCTTTATCTTGCCAACATGTGA[C/T]GCCTGGGGGTGGCTCTGTGGCTAAC
Phenotype
MIM: 610538
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
UBE2T PubMed Links

Gene Details

Gene
UBE2T
Gene Name
ubiquitin conjugating enzyme E2 T
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001310326.1 200 UTR 5 NP_001297255.1
NM_014176.3 200 Missense Mutation ATC,GTC I22V NP_054895.1

View Full Product Details