Product Details

SNP ID
rs144057351
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:114776425 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACTGCTTTCCTCATCACTTCTCCC[A/G]TTTCACAGATTCTGTCAATCTGACT
Phenotype
MIM: 611656
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SIKE1 PubMed Links

Gene Details

Gene
SIKE1
Gene Name
suppressor of IKBKE 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001102396.1 523 Missense Mutation ACG,ATG T152M NP_001095866.1
NM_025073.2 523 Missense Mutation ACG,ATG T148M NP_079349.2

View Full Product Details