Product Details

SNP ID
rs144068158
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:46189306 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGGGCTCCTGGGGCTCCCTCCTC[A/C]TTTGGGGGTGGCTCCAGGAAAATTG
Phenotype
MIM: 606822 MIM: 613170
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
POMGNT1 PubMed Links

Gene Details

Gene
POMGNT1
Gene Name
protein O-linked mannose N-acetylglucosaminyltransferase 1 (beta 1,2-)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001243766.1 1985 Nonsense Mutation GGA,TGA G641* NP_001230695.1
NM_001290129.1 1985 Missense Mutation AAG,AAT K627N NP_001277058.1
NM_001290130.1 1985 Missense Mutation AAG,AAT K506N NP_001277059.1
NM_017739.3 1985 Missense Mutation AAG,AAT K649N NP_060209.3
XM_005271010.1 1985 Intron XP_005271067.1
XM_006710755.1 1985 Intron XP_006710818.1
XM_006710756.1 1985 Nonsense Mutation GGA,TGA G641* XP_006710819.1
XM_011541760.2 1985 Missense Mutation AAG,AAT K627N XP_011540062.1
XM_017001690.1 1985 Missense Mutation AAG,AAT K649N XP_016857179.1
Gene
TSPAN1
Gene Name
tetraspanin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005727.3 1985 Intron NP_005718.2
XM_011540460.2 1985 Intron XP_011538762.1

View Full Product Details