Product Details

SNP ID
rs144092881
Assay Type
Functionally tested
NCBI dbSNP Submissions
7
Location
Chr.1:53088958 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGCCAGCGCATCACCCAGCACGTT[A/G]ATCATGGTGCGGAAACGGTCCCTGG
Phenotype
MIM: 608661 MIM: 604471
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PODN PubMed Links

Gene Details

Gene
PODN
Gene Name
podocan
There are no transcripts associated with this gene.

Gene
SLC1A7
Gene Name
solute carrier family 1 member 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001287595.1 1246 Silent Mutation ATC,ATT I520I NP_001274524.1
NM_001287596.1 1246 Intron NP_001274525.1
NM_001287597.1 1246 Silent Mutation ATC,ATT I389I NP_001274526.1
NM_006671.5 1246 Silent Mutation ATC,ATT I461I NP_006662.3
XM_011542002.2 1246 Silent Mutation ATC,ATT I238I XP_011540304.1

View Full Product Details