Product Details

SNP ID
rs144106336
Assay Type
Functionally tested
NCBI dbSNP Submissions
6
Location
Chr.1:11273813 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTGTGCCGTGGCTGTCCTGGCTGT[G/T]CACGGGGCCGGTAATTTGGTCAACA
Phenotype
MIM: 611632
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
UBIAD1 PubMed Links

Gene Details

Gene
UBIAD1
Gene Name
UbiA prenyltransferase domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013319.2 616 Silent Mutation GTG,GTT V94V NP_037451.1
XM_006710590.3 616 Silent Mutation GTG,GTT V94V XP_006710653.1
XM_011541304.2 616 Silent Mutation GTG,GTT V94V XP_011539606.1

View Full Product Details