Product Details

SNP ID
rs144116304
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:161099761 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAGAAACTCTGACCAGAGCTAGAG[A/T]CACCATCTGCAATGATCTCTACATC
Phenotype
MIM: 613466
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
KLHDC9 PubMed Links

Gene Details

Gene
KLHDC9
Gene Name
kelch domain containing 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001007255.2 996 UTR 3 NP_001007256.1
NM_152366.4 996 Missense Mutation GAC,GTC D284V NP_689579.3
Gene
PFDN2
Gene Name
prefoldin subunit 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012394.3 996 Intron NP_036526.2
XM_011509624.2 996 Intron XP_011507926.1

View Full Product Details