Product Details

SNP ID
rs144119137
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:54986160 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGCATGACAGTCAGCGGCACAGTG[A/G]TTCTGGTGGCCGGGACGCTCTGCTT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TMEM61 PubMed Links

Gene Details

Gene
TMEM61
Gene Name
transmembrane protein 61
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_182532.2 481 Missense Mutation ATT,GTT I27V NP_872338.1
XM_005270586.4 481 Missense Mutation ATT,GTT I22V XP_005270643.1
XM_011540911.2 481 Missense Mutation ATT,GTT I27V XP_011539213.1
XM_011540912.2 481 Missense Mutation ATT,GTT I22V XP_011539214.1

View Full Product Details