Product Details

SNP ID
rs144136736
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:153302432 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTGATATCCAATGTCACAAAAGTT[A/C]CGTGTGTCCATGTGAAAGGACTGTA
Phenotype
MIM: 608197
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
PGLYRP3 PubMed Links

Gene Details

Gene
PGLYRP3
Gene Name
peptidoglycan recognition protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_052891.2 1187 Silent Mutation CGG,CGT R235R NP_443123.1
XM_011509118.1 1187 Silent Mutation CGG,CGT R271R XP_011507420.1
XM_011509119.1 1187 Silent Mutation CGG,CGT R235R XP_011507421.1
XM_011509120.2 1187 Silent Mutation CGG,CGT R233R XP_011507422.1

View Full Product Details