Product Details

SNP ID
rs144211111
Assay Type
Functionally tested
NCBI dbSNP Submissions
12
Location
Chr.1:155257587 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAGGATAGAAAGCCCAAAGCCTGC[A/G]CCATTGTTGGTTTCCACACAGAAGC
Phenotype
MIM: 602989 MIM: 606913
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CLK2 PubMed Links

Gene Details

Gene
CLK2
Gene Name
CDC like kinase 2
There are no transcripts associated with this gene.

Gene
FAM189B
Gene Name
family with sequence similarity 189 member B
There are no transcripts associated with this gene.

Gene
SCAMP3
Gene Name
secretory carrier membrane protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005698.3 744 Silent Mutation NP_005689.2
NM_052837.2 744 Silent Mutation NP_443069.1
XM_006711105.3 744 Silent Mutation XP_006711168.1
XM_006711106.3 744 Silent Mutation XP_006711169.1
XM_016999991.1 744 Intron XP_016855480.1

View Full Product Details