Product Details

SNP ID
rs144230559
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:117113386 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCATGGTGCCTCCACAGGCGCAGC[A/G]AGCGGTTTTCTGGCCCCCGCTGGAG
Phenotype
MIM: 609318 MIM: 604718
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TRIM45 PubMed Links

Gene Details

Gene
TRIM45
Gene Name
tripartite motif containing 45
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145635.1 2155 Missense Mutation CGC,TGC R505C NP_001139107.1
NM_025188.3 2155 Missense Mutation CGC,TGC R523C NP_079464.2
XM_011542199.2 2155 Missense Mutation CGC,TGC R419C XP_011540501.1
Gene
TTF2
Gene Name
transcription termination factor 2
There are no transcripts associated with this gene.

View Full Product Details