Product Details

SNP ID
rs144252979
Assay Type
Functionally tested
NCBI dbSNP Submissions
12
Location
Chr.1:210339736 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTCACATGAGTGACCTCAGGAGGG[A/T]CATGAACAGAACTGTCCAGACTGCC
Phenotype
MIM: 605743
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
HHAT PubMed Links

Gene Details

Gene
HHAT
Gene Name
hedgehog acyltransferase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001122834.3 Intron NP_001116306.1
NM_001170564.2 Intron NP_001164035.1
NM_001170580.2 Intron NP_001164051.1
NM_001170587.2 Intron NP_001164058.1
NM_001170588.2 Intron NP_001164059.1
NM_018194.5 Intron NP_060664.2
XM_006711441.3 Intron XP_006711504.1
XM_011509737.2 Intron XP_011508039.1
XM_011509738.2 Intron XP_011508040.1
XM_011509739.2 Intron XP_011508041.1
XM_011509740.2 Intron XP_011508042.1
XM_011509741.2 Intron XP_011508043.1
XM_011509742.2 Intron XP_011508044.1
XM_011509744.2 Intron XP_011508046.1
XM_011509746.2 Intron XP_011508048.1
XM_011509747.1 Intron XP_011508049.1
XM_017001726.1 Intron XP_016857215.1
XM_017001727.1 Intron XP_016857216.1
XM_017001728.1 Intron XP_016857217.1
XM_017001729.1 Intron XP_016857218.1
XM_017001730.1 Intron XP_016857219.1
XM_017001731.1 Intron XP_016857220.1
XM_017001732.1 Intron XP_016857221.1
XM_017001733.1 Intron XP_016857222.1
XM_017001734.1 Intron XP_016857223.1
XM_017001735.1 Intron XP_016857224.1
XM_017001736.1 Intron XP_016857225.1
XM_017001737.1 Intron XP_016857226.1
XM_017001738.1 Intron XP_016857227.1
XM_017001739.1 Intron XP_016857228.1
XM_017001740.1 Intron XP_016857229.1
XM_017001741.1 Intron XP_016857230.1
XM_017001742.1 Intron XP_016857231.1
XM_017001743.1 Intron XP_016857232.1
XM_017001744.1 Intron XP_016857233.1

View Full Product Details