Product Details

SNP ID
rs144324004
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:75919092 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTAATAGTTCACATGGATCTTTGT[A/G]TCTTGTTGAAGGAATGTAATCAAAC
Phenotype
MIM: 602105
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ASB17 PubMed Links

Gene Details

Gene
ASB17
Gene Name
ankyrin repeat and SOCS box containing 17
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_080868.2 888 Missense Mutation CAC,TAC H250Y NP_543144.1
XM_006710345.3 888 Missense Mutation CAC,TAC H202Y XP_006710408.1
XM_006710346.3 888 Missense Mutation CAC,TAC H172Y XP_006710409.1
Gene
MSH4
Gene Name
mutS homolog 4
There are no transcripts associated with this gene.

View Full Product Details